Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Arsenal™ 5008 Fire and Safety Gear Bag

Catalog No. 19819717 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Red Camo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-717 Red Camo
19-819-718 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-819-717 Supplier Ergodyne™ Supplier No. 13008

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Large capacity gear bags designed to store gear and safety equipment for fire fighters, EMTs, and tower climbers.

  • Tarpaulin reinforced bottom with molded feet for durability
  • 360 degree grab handles for handling from any angle
  • Reflective strips around the entire bag for better visibility
  • Large, 8.5 x 16 x 15.5 in. (22 x 41 x 39 cm), side pocket to separate equipment
  • Two side accessory pockets to store personal items
  • Four V-Rings to clip on SCBA mask bag, carabiner, or other items
  • ID window for user identification

Specifications

Color Red Camo
Material 1000D Polyester
Dimensions (L x W x H) 31 x 16 x 15.5 in. (79 x 41 x 39 cm)
For Use With (Application) Firefighters, EMTs, tower climbers
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.