Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Arsenal™ 5020 Standard Polyester Gear Duffel Bag

Catalog No. 19819808 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Realtree Xtra™
Volume (Metric):
24 L
54 L
79 L
112 L
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric)
19-819-808 Black 79 L
19-819-806 Black 24 L
19-819-807 Black 54 L
19-819-812 Black 112 L
19-819-826 Realtree Xtra™ 24 L
19-819-827 Realtree Xtra™ 54 L
19-819-828 Realtree Xtra™ 79 L
19-819-829 Realtree Xtra™ 112 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-819-808 Supplier Ergodyne™ Supplier No. 13321
Add to Cart
Add to Cart

Arsenal™ 5020 Standard Gear Duffel Bag is designed to store, organize, and easily carry everything you need for the workday or weekend.

  • Durable material withstands wear and tear
  • Water-resistant backing helps to repel water and stains
  • Heavy-duty webbing straps on top and side of duffel for extra reinforcement
  • Removable and adjustable padded shoulder strap for easy transportation
  • Heavy-duty zippers to secure gear inside the duffel
  • Two end accessory pockets for cell phones, headphones, pens, keys, and other small parts

Specifications

Color Black
Material 600D Polyester
Dimensions (L x W x H) 28.5 x 13 x 13 in. (73 x 33 x 33 cm)
For Use With (Application) Fire and Rescue, EMTs, Construction, General Purpose
Quantity 1 Ea.
Size Medium
Volume (Metric) 79 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.