Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Arsenal™ 5093 Quick Adjust Reflective Suspenders

Catalog No. 19819819 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-819-819 Medium
19-819-818 Small
19-819-821 Large
19-819-822 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-819-819 Supplier Ergodyne™ Supplier No. 13350

Quick Adjust Suspenders are designed to be quickly attached and feature shoulder padding for comfort.

  • Heavy-duty polyester webbing
  • H-shaped back design
  • Foam padded shoulder straps
  • Reflective trim through center

Specifications

Color Red
Size Medium
For Use With (Application) Firefighters, EMTs, Biochem, Hazmat
Length (English) Body 36 in.
Length (Metric) Body 91 cm
Material 600D Nylon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.