Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Arsenal™ 5125 Carry-On Luggage

Catalog No. 19819771 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-771 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-771 Supplier Ergodyne™ Supplier No. 13125

Arsenal™ 5125 Carry-On Luggage includes a telescoping pull handle and wheels.

  • 1680D ballistic polyester
  • YKK zippers
  • Haul handles located on the bag for easy carry
  • Molded wheels for safe and easy transport
  • Molded retractable pull handle
  • Rugged construction with extra bartacks and nylon stitching

Specifications

Color Black
Material 1680D Ballistic Polyester
Dimensions (L x W x H) 14 x 9 x 22.5 in. (36 x 23 x 57 cm)
For Use With (Application) Contractors, Trades, Public Safety, Construction, Electricians, General Purpose
Quantity 1 Ea.
Volume (Metric) 46 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.