Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenazo Iii, Solstice

Catalog No. 6000358 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-003-58 5 g
60-003-57 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-003-58 Supplier Solstice Supplier No. 110905G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for spectrophotometric det. of Th, Zr, U, Cd, Zn, Ca

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1668-00-4
Molecular Formula C22H18As2N4O14S2
Molecular Weight (g/mol) 776.363
MDL Number MFCD00036695
InChI Key TVMZRHVOFZTNET-RIRMOVKSSA-N
PubChem CID 9810878
IUPAC Name (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
SMILES C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O

Specifications

Packaging Glass Bottle
Linear Formula (HO)2C10H2(SO3H)2(N=NC6H4AsO3H2)2
UN Number 3465
Beilstein 5717957
Quantity 5 g
Formula Weight 776.37g/mol
Chemical Name or Material Arsenazo Iii
Hazard Statement P261-P273-P301 + P310-P311-P501
Precautionary Statement H301 + H331-H410
EINECSNumber 216-788-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.