CAS RN 1668-00-4
Arsenazo III Indicator, Certified, LabChem™
CAS: 1668-00-4 Molecular Formula: C22H18As2N4O14S2 Molecular Weight (g/mol): 776.363 InChI Key: TVMZRHVOFZTNET-RIRMOVKSSA-N PubChem CID: 9810878 IUPAC Name: (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid SMILES: C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O
Arsenazo II, free acid, >80%, MP Biomedicals™
CAS: 1668-00-4 Molecular Formula: C22H18As2N4O14S2 Molecular Weight (g/mol): 776.363 MDL Number: MFCD00036695 InChI Key: TVMZRHVOFZTNET-RIRMOVKSSA-N Synonym: 2,2'-[1, 8-dihydroxy-3,6-disulfo-2, 7-naphthalene-bis(azo)dibenzenearsonic acid,2, 2'-[1, 8-dihydroxy-3 PubChem CID: 9810878 IUPAC Name: (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid SMILES: C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O
Arsenazo III
CAS: 1668-00-4 Molecular Formula: C22H18As2N4O14S2 Molecular Weight (g/mol): 776.363 MDL Number: MFCD00036695 InChI Key: TVMZRHVOFZTNET-RIRMOVKSSA-N Synonym: arsenazo iii,arsenazo iii sodium salt,2,2'-1, 8-dihydroxy-3,6-disulfo-2, 7-naphthalene-bis azo dibenzenearsonic acid,2,7-bis 2-arsonophenylazo-1, 8-dihydroxy-3, 6-naphthalene disulfonic acid,2,7-bis 2-arsonophenylazo-1,8-dihydroxynaphthalene-3,6-disulfonic acid,2,7-bis 2-arsonophenylazo chromotropic acid,2,2'-1,8-dihydroxy-3,6-disulfonaphthylene-2,7-bisazo bisbenzenearsonic acid,3,6-bis 2-2-arsonophenyl diazen-1-yl-4,5-dihydroxynaphthalene-2,7-disulfonic acid,3,6-bis 2-2-arsonophenyl diazenyl-4,5-dihydroxy-2,7-naphthalenedisulfonic acid,3,6-bis 2-2-arsonophenyl hydrazin-1-ylidene-4,5-dioxonaphthalene-2,7-disulfonic acid PubChem CID: 9810878 IUPAC Name: (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid SMILES: C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O
Arsenazo III, Sigma-Aldrich
CAS: 1668-00-4 Molecular Weight (g/mol): 776.37 g/mol
Arsenazo III 95.0+%, TCI America™
CAS: 1668-00-4 Molecular Formula: C22H18As2N4O14S2 Molecular Weight (g/mol): 776.363 MDL Number: MFCD00036695 InChI Key: TVMZRHVOFZTNET-RIRMOVKSSA-N Synonym: arsenazo iii,arsenazo iii sodium salt,2,2'-1, 8-dihydroxy-3,6-disulfo-2, 7-naphthalene-bis azo dibenzenearsonic acid,2,7-bis 2-arsonophenylazo-1, 8-dihydroxy-3, 6-naphthalene disulfonic acid,2,7-bis 2-arsonophenylazo-1,8-dihydroxynaphthalene-3,6-disulfonic acid,2,7-bis 2-arsonophenylazo chromotropic acid,2,2'-1,8-dihydroxy-3,6-disulfonaphthylene-2,7-bisazo bisbenzenearsonic acid,3,6-bis 2-2-arsonophenyl diazen-1-yl-4,5-dihydroxynaphthalene-2,7-disulfonic acid,3,6-bis 2-2-arsonophenyl diazenyl-4,5-dihydroxy-2,7-naphthalenedisulfonic acid,3,6-bis 2-2-arsonophenyl hydrazin-1-ylidene-4,5-dioxonaphthalene-2,7-disulfonic acid PubChem CID: 9810878 IUPAC Name: (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid SMILES: C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O
Arsenazo III, 0.2% (w/v), Ricca Chemical
CAS: 1668-00-4 Molecular Formula: C22H18As2N4O14S2 Molecular Weight (g/mol): Mixture InChI Key: TVMZRHVOFZTNET-RIRMOVKSSA-N PubChem CID: 9810878 IUPAC Name: (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid SMILES: C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O
Arsenazo Iii, Solstice
CAS: 1668-00-4 Molecular Formula: C22H18As2N4O14S2 Molecular Weight (g/mol): 776.363 MDL Number: MFCD00036695 InChI Key: TVMZRHVOFZTNET-RIRMOVKSSA-N PubChem CID: 9810878 IUPAC Name: (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid SMILES: C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O
Arsenazo-lll, free acid, 80.85%, MP Biomedicals™
CAS: 1668-00-4 Molecular Formula: C22H18As2N4O14S2 Molecular Weight (g/mol): 776.363 InChI Key: TVMZRHVOFZTNET-RIRMOVKSSA-N Synonym: 2,7-bis(2-arsonophenylazo)-1, 8-dihydroxy-3, 6-naphthalene disulfonic acid,2, 7-bis(2-arsonophenylazo)-1, 8-dihydroxy-3 PubChem CID: 9810878 IUPAC Name: (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid SMILES: C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O