Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenazo III, Sigma-Aldrich

Catalog No. 01259028 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-028 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-028 Supplier MilliporeSigma Supelco Supplier No. A9277510G
Add to Cart
Add to Cart

Arsenazo III is a metallochromic dye. Arsenazo III is used in the determination of calcium in biological samples. It is used to evaluate calcium transport in permeabilized cells. It is also used for the detection of rare earth metals (polyvalent metal ions). Arsenazo III is used for the spectrophotometric determination of uranium and thorium.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1668-00-4
Molecular Weight (g/mol) 776.37 g/mol

Specifications

Color Black to Brown
Quantity 10 g
Chemical Name or Material Arsenazo III
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.