Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenazo-lll, free acid, 80.85%, MP Biomedicals™

Catalog No. ICN19505905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19505905 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19505905 Supplier MP Biomedicals, Inc Supplier No. 0219505905

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1668-00-4
Molecular Formula C22H18As2N4O14S2
Molecular Weight (g/mol) 776.363
InChI Key TVMZRHVOFZTNET-RIRMOVKSSA-N
Synonym 2,7-bis(2-arsonophenylazo)-1, 8-dihydroxy-3, 6-naphthalene disulfonic acid, 2, 7-bis(2-arsonophenylazo)-1, 8-dihydroxy-3
PubChem CID 9810878
IUPAC Name (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
SMILES C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O

Specifications

Color Brown
Quantity 5 g
Physical Form Powder
Formula Weight 776.04
Percent Purity 80.85%
Chemical Name or Material Arsenazo-lll
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.