Learn More
Arsenic(III) sulfide, 99.999% (metals basis)
Description
Arsenic(III) sulfide is used to make infrared-transmitting glass, pyrotechnics, pigments, linoleum, electrical semiconductors and photoconductors. It acts as a reducing agent and serves as a tanning agent. It is also used for depilating hides and veterinary medication. Further, it is used as an inorganic photoresist to fabricate 3-D nanostructures. In addition to this, it is used in the treatment for acute promyelocytic leukemia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 707°C |
| Melting Point | 300°C |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99.99% (Metals basis) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 246.03 |
| Density | 3.43 g/cm3 |
| Chemical Name or Material | Arsenic(III) sulfide |
Safety and Handling
| Hazard Category | H301+H331-H350 |
| Hazard Statement | When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c |
| DOTInformation | DOT Class: 6.1, Packaging Group: III |
| EINECSNumber | 215-117-4 |
| RTECSNumber | CG2638000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.