Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic Trioxide, Powder, ACS, 99.95-100.05%, Spectrum™ Chemical
SDP

Catalog No. 18608938 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
125 g
500 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-938 5 g Amber Glass Bottle
18-608-934 25 g Amber Glass Bottle
18-608-936 125 g Amber Glass Bottle
18-608-937 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-608-938 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A13405GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1327-53-3
Molecular Formula As2O3
Molecular Weight (g/mol) 197.84
MDL Number MFCD00003433
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
IUPAC Name diarsorosooxidane
SMILES O=[As]O[As]=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 5 g
Percent Purity 99.95 to 100.05%
Grade Reagent
Packaging Amber Glass Bottle
Residue after Ignition 0.0002
Chemical Name or Material Arsenic Trioxide, Powder
Hazard Category c
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.