accessibility menu, dialog, popup

UserName

Viewing profile as user:
OAsOAsO

CAS RN 1327-53-3

IUPAC Name: diarsorosooxidane
Synonyms: arsenic trioxide
Molecular Weight (g/mol): 197.84
Molecular Formula: As2O3
InChi Key: IKWTVSLWAPBBKU-UHFFFAOYSA-N
SMILES: O=[As]O[As]=O
Molecular Weight (g/mol) 
  • (29)
  • (2)
Grade 
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (5)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (4)
  • (12)
Percent Purity 
  • (1)
  • (1)
  • (3)
  • (6)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (9)
  • (2)
  • (4)
  • (11)
  • (29)
  • (2)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (12)
  • (1)
  • (1)
  • (3)
  • (6)
  • (3)
  • (2)
  • (1)