Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic Trioxide, Reagent Grade, ≥ 99.0%, LabChem™

Catalog No. LC115059 Shop All LabChem Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC115059 100 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC115059 Supplier LabChem, Inc. Supplier No. LC115059
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1327-53-3
Molecular Formula As2O3
Molecular Weight (g/mol) 197.84
MDL Number MFCD00003433
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
IUPAC Name diarsorosooxidane
SMILES O=[As]O[As]=O

Specifications

Boiling Point 102°C
Melting Point -7°C
Color White
CAS Max % 100
Physical Form Powder
Quantity 100 g
Linear Formula As2O3
UN Number UN1561
Solubility Information Soluble in water
Formula Weight 197.84
Percent Purity ≥99.0%
Grade Reagent
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1.02g/mL
Decomposition Information Arsenic and its oxides
Density 1.02g/mL
Chemical Name or Material Arsenic Trioxide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Causes severe skin burns and eye damage.
May cause cancer.
Toxic to aquatic life.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe dust.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
EINECSNumber 215-481-4
Recommended Storage Room Temperature
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.