Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenite Standard, 0.100 N (N/10), Ricca Chemical
SDP

Catalog No. 82032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
820-32 1 L Natural Poly Bottle
820-16 500 mL Natural Poly Bottle
820-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 820-32 Supplier Ricca Chemical Company Supplier No. 82032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1327-53-3 , 144-55-8 , 1310-73-2 , 7732-18-5
Molecular Formula As2O3
Molecular Weight (g/mol) 197.84
MDL Number MFCD00003433
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
IUPAC Name diarsorosooxidane
SMILES O=[As]O[As]=O

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 0.100Normal
CAS Min % 97.0, 0.48, 0.39, 0.21
CAS Max % 100.0, 0.5, 0.4, 0.22
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 1 L
Chemical Name or Material Arsenite Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.