Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Barrier Pipette Tips in Bulk Packaging

Catalog No. 2140280
Change view
Click to view available options
Volume (Metric):
200 μL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-402-80 200 μL
21-236-29 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-402-80 Supplier Thermo Scientific™ Supplier No. 2069B

Thermo Scientific™ ART™ Barrier Pipette tips seal in contaminants, providing bi-directional protection for samples and pipettes. Fits most pipettes.

ART™ Barrier tips are not the same as traditional filter pipette tips. ART Barrier tips contain a one-of-a-kind, self-sealing barrier that seals in aerosol and liquid contaminants on contact, providing bi-directional protection from contamination for pipettes and samples.

  • Universal Tip, fits most brands of pipettes
  • ART™ tips are certified to be free DNase, RNase, DNA, ATP, Endotoxin, and Pyrogens
  • Convenient bulk packaging for high volume usage

  • Sterile
  • Low Retention
  • Racked with lift-off lid
TRUSTED_SUSTAINABILITY

Specifications

Description ART 200, Filtered, Non-sterile, Bulk
Volume (Metric) 200 μL
Filtered Filtered
Sterility Non-sterile
Color Clear
Rack Color bulk - no rack
Autoclavable Not Autoclavable
Model ART 200
Length (English) 2.32 in
Length (Metric) 5.88 cm
Clean Claims DNase, RNase, DNA, ATP, Endotoxin, and Pyrogen Free
Features Self-sealing ART Barrier
Format Bulk
Tip Style Micro Point
Packaging 5 bags of 1000 tips each
Quantity 5000 tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Product Line ART
Material Virgin Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.