Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Barrier Reload Insert Pipette Tips
GREENER_CHOICE

Catalog No. 02682240
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
100 μL
200 μL
Model:
ART 10
ART 100
ART 100E
ART 200
ART 20E
ART 20P
Tip Style:
Low Retention, Micro Point
Micro Point
Fit for Purpose:
Sterile and PCR Ready
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Model Tip Style Fit for Purpose
02-682-240 10 μL ART 10 Low Retention, Micro Point Sterile and PCR Ready
02-682-238 100 μL ART 100E Low Retention, Micro Point Sterile and PCR Ready
02-682-237 100 μL ART 100E Micro Point Sterile and PCR Ready
02-682-232 100 μL ART 100 Micro Point Sterile and PCR Ready
02-682-231 200 μL ART 200 Low Retention, Micro Point Sterile and PCR Ready
02-682-230 200 μL ART 200 Micro Point Sterile and PCR Ready
02-682-239 10 μL ART 10 Micro Point Sterile and PCR Ready
02-682-241 10 μL ART 20E Micro Point Sterile and PCR Ready
02-682-236 20 μL ART 20P Low Retention, Micro Point Sterile and PCR Ready
02-682-235 20 μL ART 20P Micro Point Sterile and PCR Ready
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 02-682-240 Supplier Thermo Scientific™ Supplier No. 213905RI
Add to Cart
Add to Cart

Obtain accurate and consistent performance with these pipette tips, which seal when exposed to potential contaminants.

Obtain accurate and consistent performance with Thermo Scientific™ ART™ Barrier Reload Insert Pipette Tips, which seal when exposed to potential contaminants. The ART inserts are environmentally friendly, providing individually packaged blister tubs that are made out of recycled plastic, designed to be used with the reusable ART hinged racks and creating the ART Reload System. The tips from the reload inserts clip securely into the hinged racks for efficient reloading.

Features

  • Sterile tips certified free from RNase, DNase and pyrogen
  • Available in MicroPoint, extended length, low retention and SoftFit-L™ designed for use with Rainin™ LTS™ LiteTouch™ Pipettes

Recommended for:

  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Filtered Filtered
Sterility Sterile
Model ART 10
Length (English) 1.23 in.
Length (Metric) 3.13cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Refill
Tip Style Low Retention, Micro Point
For Use With (Equipment) Universal Fit
Product Line ART
Fit for Purpose Sterile and PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.