Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Barrier Reload Insert, Extended Length Pipette Tips

Catalog No. 02682242
Change view
Click to view available options
Volume (Metric):
10 μL
1000 μL
Tip Style:
Low Retention
Low Retention, Extended Length
Micro Point, Extended Length
Micro Point, Extended Length, Low Retention
Micro Point, Low Retention
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Tip Style
02-682-242 10 μL Micro Point, Low Retention
02-682-234 10 μL Micro Point, Extended Length, Low Retention
02-682-233 10 μL Micro Point, Extended Length
02-682-244 1000 μL Low Retention
02-682-243 1000 μL Low Retention, Extended Length
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-682-242 Supplier Thermo Scientific™ Supplier No. 2149E05RI

Obtain accurate and consistent performance with these pipette tips, which seal when exposed to potential contaminants.

Obtain accurate and consistent performance with Thermo Scientific™ ART™ Barrier Reload Insert Pipette Tips, which seal when exposed to potential contaminants. The ART inserts are environmentally friendly, providing individually packaged blister tubs that are made out of recycled plastic, designed to be used with the reusable ART hinged racks and creating the ART Reload System. The tips from the reload inserts clip securely into the hinged racks for efficient reloading.

Features

  • Sterile tips certified free from RNase, DNase and pyrogen
  • Available in MicroPoint,extended length, low retention and SoftFit-L™ designed for use with Rainin™ LTS™ LiteTouch™ Pipettes

Recommended for:

  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
TRUSTED_SUSTAINABILITY

Specifications

Description ART 20E, Filtered, Sterile, Refill
Volume (Metric) 10 μL
Filtered Filtered
Sterility Sterile
Model ART 20E
Length (English) 1.8 in.
Length (Metric) 4.57cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Refill
Tip Style Micro Point, Low Retention
For Use With (Equipment) Universal Fit
Product Line ART
Fit for Purpose Sterile & PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.