Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Barrier Specialty Pipette Tips

Catalog No. 21402649
Change view
Click to view available options
Volume (Metric):
1250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-402-649 1250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-402-649 Supplier Thermo Scientific™ Supplier No. 2080B
This item is not returnable. View return policy

Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.

Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to aid in pipetteting for a variety of applications:

Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel

Gel loading for loading acrylamide or agarose gels

Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials

Solvent Safe for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettetes

Ergonomic pipette tips are designed for Gilson pipetteman™ pipettetes and Thermo Scientific™ Finnpipettete™ Focus pipettetes

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1250 μL
Sterility Non-sterile
Model ART 1250
Length (English) 4.033 in.
Length (Metric) 10.244cm
Format Bulk
Tip Style Extended Length, Ergonomic
For Use With (Equipment) Gilson Pipetman, Thermo Scientific Finnpipette Focus
Product Line ART
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.