Learn More
Thermo Scientific™ ART™ Non-Filtered Extended-Length Wide-Bore Genomic pipette Tips
GREENER_CHOICE
Catalog No. 21402157
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Tip Style:
Extended Length
Genomic/Wide Orifice, Extended Length
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Tip Style
21-402-157 Sterile Extended Length
21-402-158 Non-sterile Genomic/Wide Orifice, Extended Length
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-402-157 Supplier Thermo Scientific™ Supplier No. 3701
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Ideal for very viscous material
These specialty Wide Bore Tips are designed for researchers dealing with macromolecules like genomic DNA and are especially critical when transferring fragile cellular samples such as macrophages, hybridomas and hepatocytes. When these types of samples are forced through the narrow tip orifice of conventional tips, they are subjected to mechanical shearing forces that can cause trauma and fragmentation to the cells being transferred. The result is often a decrease in plating capability and compromised cell viability. Our wide bore blunt end tip orifice eliminates cell shearing and flow resistance, making this tip ideal for very viscous material.
  • Clear, extended-length tips with large orifice
  • For working with macromolecules, viscous material, plant tissues, or doing cell suspensions
  • Extended length is perfect for those hard-to-reach applications
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Clean Claims DNase, RNase, DNA, ATP and Endotoxin Free
Format Rack
Tip Style Extended Length
Product Line ART
Fit for Purpose Sterile