Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Non-Filtered Standard Hinged Rack Pipette Tips
GREENER_CHOICE

Catalog No. 13811156
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
200 μL
300 μL
1000 μL
Tip Style:
Extended Length
Low Retention
Standard
For Use With (Equipment):
Eppendorf
Universal Fit
Fit for Purpose:
Certified Quality
Sterile
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Tip Style For Use With (Equipment) Fit for Purpose
13-811-156 1000 μL Standard Universal Fit Certified Quality
13-811-159 1000 μL Low Retention Universal Fit Sterile
13-811-158 1000 μL Low Retention Universal Fit Certified Quality
13-811-157 1000 μL Standard Universal Fit Sterile
13-811-147 300 μL Low Retention Universal Fit Sterile
13-811-146 300 μL Low Retention Universal Fit Certified Quality
13-811-144 300 μL Standard Universal Fit Certified Quality
13-811-145 300 μL Standard Universal Fit Sterile
13-811-129 200 μL Low Retention Universal Fit Sterile
13-811-128 200 μL Low Retention Universal Fit Certified Quality
13-811-127 200 μL Standard Universal Fit Sterile
13-811-126 200 μL Standard Universal Fit Certified Quality
13-811-119 20 μL Low Retention Eppendorf Sterile
13-811-118 20 μL Low Retention Eppendorf Certified Quality
13-811-116 20 μL Standard Eppendorf Certified Quality
13-811-111 10 μL Extended Length Universal Fit Sterile
13-811-105 10 μL Low Retention Universal Fit Sterile
13-811-104 10 μL Low Retention Universal Fit Certified Quality
13-811-103 10 μL Standard Universal Fit Sterile
13-811-102 10 μL Standard Universal Fit Certified Quality
13-811-117 20 μL Standard Eppendorf Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Catalog No. 13-811-156 Supplier Thermo Scientific™ Supplier No. 3102HR
Add to Cart
Add to Cart

Access tips one-handed with this versatile hinged rack, which is designed for the way you work.

Access tips with one hand using Thermo ScientificTM ARTTM Nonfiltered Hinged Rack Pipette Tips, which is designed for the way you work. No longer worry about your tips spilling on your bench or floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover--now you have a traditional lift-off when you need it.

Features:

  • Complete selection of non-filtered and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
TRUSTED_SUSTAINABILITY

Specifications

Description ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack
Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Model ART Non-filtered
Length (English) 3.5 in.
Length (Metric) 8.89 cm
Clean Claims DNase, RNase, DNA, ATP and Endotoxin Free
Format Hinged Rack
Tip Style Standard
For Use With (Equipment) Universal Fit
Product Line ART
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.