Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Non-Filtered Extended Length Hinged Rack Pipette Tips
GREENER_CHOICE

Catalog No. 13811135
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
1000 μL
1250 μL
Sterility:
Non-sterile
Sterile
Tip Style:
Low Retention
Universal Fit
Fit for Purpose:
Certified Quality
Sterile
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Tip Style Fit for Purpose
13-811-135 200 μL Sterile Universal Fit Sterile
13-811-110 10 μL Non-sterile Universal Fit Certified Quality
13-811-112 10 μL Non-sterile Low Retention Certified Quality
13-811-113 10 μL Sterile Low Retention Sterile
13-811-134 200 μL Non-sterile Universal Fit Certified Quality
13-811-136 200 μL Non-sterile Low Retention Certified Quality
13-811-137 200 μL Sterile Low Retention Sterile
13-811-166 1000 μL Non-sterile Universal Fit Certified Quality
13-811-168 1000 μL Non-sterile Low Retention Certified Quality
13-811-167 1000 μL Sterile Universal Fit Sterile
13-811-169 1000 μL Sterile Low Retention Sterile
13-811-172 1250 μL Non-sterile Universal Fit Certified Quality
13-811-173 1250 μL Sterile Universal Fit Sterile
13-811-174 1250 μL Non-sterile Low Retention Certified Quality
13-811-175 1250 μL Sterile Low Retention Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 13-811-135 Supplier Thermo Scientific™ Supplier No. 3541HR
Add to Cart
Add to Cart

Access tips one-handed with this versatile hinged rack, designed for the way you work.

Access tips with one hand using Thermo Scientific™ ART™ Non-Filtered Extended Length Hinged Rack Pipette Tips, designed for the way you work. No longer worry about your tips spilling on your bench or the floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover — now you have a traditional lift-off when you need it.

Features:

  • Complete selection of non-filtered and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
TRUSTED_SUSTAINABILITY

Specifications

Description ART Non-Filtered, Sterile, Rack
Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Natural
Model ART Non-filtered
Length (English) 3.6 in.
Length (Metric) 9.09cm
Clean Claims DNase, RNase, DNA, ATP and Endotoxin Free
Format Rack
Tip Style Universal Fit
For Use With (Equipment) Universal Fit
Product Line ART
Fit for Purpose Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.