Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Non-filtered Low Retention Pipette Tips in Lift-off Lid Rack
GREENER_CHOICE

Catalog No. 21402498
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
300 μL
Sterility:
Non-sterile
Sterile
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility
21-402-498 10 μL Non-sterile
21-402-507 300 μL Sterile
21-402-508 300 μL Non-sterile
21-402-497 10 μL Sterile
21-402-494 20 μL Sterile
21-402-495 20 μL Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 21-402-498 Supplier Thermo Scientific™ Supplier No. 351205
Add to Cart
Add to Cart

Specifically designed for eliminating sample retention within the pipette tip.

Thermo Scientific™ ART™ Non-filtered Low Retention Pipette Tips in Lift-off Lid Rack are specifically designed for eliminating sample retention within the pipette tip.

  • Universal Tip, fits most brands of pipettes
  • ART™ non-filtered tips deliver the same proven quality, performance and reliability as ART™ barrier
  • Non-filtered tip
  • ART™ tips are certified to be free of DNA, RNase, DNase, ATP, Endotoxins, and Pyr
  • Lift-off lid racks provide access to all tips, great for multichannel
  • Proprietary low retention technology maximizes sample delivery
TRUSTED_SUSTAINABILITY

Specifications

Description ART 10 REACH, Non-filtered, Non-sterile, Racked, Lift-off Lid
Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model ART 10 REACH
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor, Endotoxin, and Pyrogen Free
Format Lift-off Lid Rack
Tip Style Low Retention
Packaging 96 tips/rack
Quantity Case of 4800
Disposable Single-use
For Use With (Equipment) Universal Fit
Product Line ART
Material Virgin Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.