Learn More
Thermo Scientific™ ART™ Non-Filtered Low Retention Universal Pipette Tips
GREENER_CHOICE
Catalog No. 21403519
Change view
Click to view available options
Volume (Metric):
0.5 to 200 μL
100 to 1000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-403-519 0.5 to 200 μL
21-403-600 100 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-403-519 Supplier Thermo Scientific™ Supplier No. 394105
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifically designed for eliminating sample retention within the pipette tip
  • Inner surface is extremely hydrophobic, increasing sample reproducibility and sample accuracy
  • Crystal clear tips allow you to easily see transparent samples
  • Reference marks on many Thermo Scientific ART tips provide a quick and convenient guide to visually establish whether you are pipetting desired volume of liquid
  • All Pure tips are racked, presterilized and certified RNase-, DNase-, DNA- and pyrogen-free
  • Other Thermo Scientific racked and bulk tips are nonsterile and autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.5 to 200 μL
Filtered Non-filtered
Sterility Sterile
Color Yellow
Autoclavable Autoclavable
Clean Claims DNase, RNase, DNA, ATP and Endotoxin Free
Format Rack
Tip Style Low Retention
Product Line ART
Fit for Purpose Sterile