Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Non-Filtered Pipette Tips, TipCycle Recycle Kit, Hinged Rack, Sterile
GREENER_CHOICE

Catalog No. 02681554
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
200 μL
1000 μL
Format:
Hinged Rack
Tip Style:
Extended Length
Standard
Fit for Purpose:
Sterile
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Format Tip Style Fit for Purpose
02-681-554 1000 μL Hinged Rack Extended Length Sterile
02-681-556 10 μL Hinged Rack Standard Sterile
02-681-560 20 μL Hinged Rack Extended Length Sterile
02-681-562 200 μL Hinged Rack Standard Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-681-554 Supplier Thermo Scientific™ Supplier No. 3101RCHR
Add to Cart
Add to Cart

The pipette tip and rack mail-back recycling program provides a simple and effective solution for recycling single use plastics. The TipCycle Recycle Kit arrives with your case of pipette tips to collect used pipette tips and empty racks. Attach the pre-paid shipping label and return for recycling.

Access tips with one hand using Thermo Scientific™ ART™ Non-filtered Hinged Rack Pipette Tips, designed for the way you work. No longer worry about your tips spilling on your bench or floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover - now you have a traditional lift-off when you need it.

  • Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
  • Complete TipCycle Pipette Tip Recycling Kit Includes: 1 case of pipette tips, box liner, liner tie, FedEx return shipping label, waiver form, and instructions for return
  • Complete selection of non-filtered and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Model ART 1000
Length (English) 3.5 in.
Length (Metric) 8.89 cm
Certifications/Compliance DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin free
Clean Claims DNase, RNase, DNA, ATP and Endotoxin Free
Format Hinged Rack
Tip Style Extended Length
Packaging 3072/Cs.
Quantity 3072/Cs.
Product Type TipCycle, Pipette Tip and Rack Recycling Kit
Includes 1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
For Use With (Equipment) Universal Fit
Product Line ART
Fit for Purpose Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.