Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Pipette Tips, TipCycle Recycle Kit, Hinged Rack, Sterile
GREENER_CHOICE

Catalog No. 02681547
Change view
Click to view available options
Volume (Metric):
10 μL
1000 μL
Tip Style:
Extended Length, Micro Point
Low Retention, Extended Length, Micro Point
Model:
ART 10 REACH
ART 1000
ART 1000L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Tip Style Model
02-681-547 1000 μL Extended Length, Micro Point ART 1000L
02-681-543 1000 μL Extended Length, Micro Point ART 1000
02-681-545 10 μL Low Retention, Extended Length, Micro Point ART 10 REACH
02-681-574 10 μL Extended Length, Micro Point ART 10 REACH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-681-547 Supplier Thermo Scientific™ Supplier No. 2179RC05HR
Add to Cart
Add to Cart

The pipette tip and rack mail-back recycling program provides a simple and effective solution for recycling single use plastics. The TipCycle Recycle Kit arrives with your case of pipette tips to collect used pipette tips and empty racks. Attach the pre-paid shipping label and return for recycling.

Unlike standard filter tips, Thermo Scientific™ ART™ Barrier Extended Length Pipette Tips in Hinged Racks seal when exposed to potential contaminants. The pipette tips from the reload inserts fit securely into the environmentally friendly, reusable hinged racks. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. Available in low retention, micropoint, extended length, and other specialty styles.

Features:

  • Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
  • Complete TipCycle Pipette Tip Recycling Kit Includes: 1 case of pipette tips, box liner, liner tie, FedEx return shipping label, waiver form, and instructions for return
  • Stackable hinged racks save space in your lab
  • Sterile tips are certified free from RNase, DNase and pyrogen

Recommended for:

  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Model ART 1000L
Length (English) 3.5 in.
Length (Metric) 8.89 cm
Certifications/Compliance Free of RNase, DNase, DNA, ATP, Bioburden, Endotoxin, Pyrogen, and PCR Inhibitors
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
For Use With (Application) Liquid Handling, Molecular Biology, PCR, Biotechnology
Format Hinged Rack
Tip Style Extended Length, Micro Point
Packaging 96 Tips/Tray, 32 Trays/Cs.
Quantity 3072 Tips/Cs.
Product Type TipCycle Recycle Kit
Green Features Recycling program for nonhazardous pipette tips and racks; environmentally friendly reusable hinged rack and reload inserts
Color Code None
For Use With (Equipment) Universal Fit
Product Line ART
Material Polypropylene
Fit for Purpose Sterile and PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.