Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Reload System
GREENER_CHOICE

Catalog No. 13811161
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
200 μL
300 μL
1000 μL
For Use With (Equipment):
Eppendorf
Universal Fit
Tip Style:
Low Retention
Standard
Fit for Purpose:
Certified Quality
Sterile
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) For Use With (Equipment) Tip Style Fit for Purpose
13-811-161 1000 μL Universal Fit Standard Sterile
13-811-163 1000 μL Universal Fit Low Retention Sterile
13-811-160 1000 μL Universal Fit Standard Certified Quality
13-811-162 1000 μL Universal Fit Low Retention Certified Quality
13-811-108 10 μL Universal Fit Low Retention Sterile
13-811-106 10 μL Universal Fit Standard Sterile
13-811-109 10 μL Universal Fit Low Retention Certified Quality
13-811-107 10 μL Universal Fit Standard Certified Quality
13-811-123 20 μL Eppendorf Low Retention Sterile
13-811-121 20 μL Eppendorf Standard Sterile
13-811-122 20 μL Eppendorf Low Retention Certified Quality
13-811-120 20 μL Eppendorf Standard Certified Quality
13-811-133 200 μL Universal Fit Low Retention Sterile
13-811-131 200 μL Universal Fit Standard Sterile
13-811-132 200 μL Universal Fit Low Retention Certified Quality
13-811-130 200 μL Universal Fit Standard Certified Quality
13-811-150 300 μL Universal Fit Low Retention Certified Quality
13-811-148 300 μL Universal Fit Standard Certified Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 13-811-161 Supplier Thermo Scientific™ Supplier No. 3101RT
Add to Cart
Add to Cart

Reduce waste and save benchtop and storage space with this advanced nonfiltered reload system.

Reduce waste and save benchtop and storage space with the advanced Thermo Scientific™ ART™ Nonfiltered Reload System. This system provides an environmentally responsible option without compromising sterility.

Features:

  • Stackable barrier tip packaging allows for tips to pass through offset holes in lower insert
  • Blister sleeve guide rails align tip inserts for smooth transfer into the rack
  • Outer sleeve protects tips from exterior contamination while refilling inserts
  • Tip packaging option reduces plastic waste
  • Rigid insert design provides reliable tip loading
  • Lot number and description are printed on every insert

Note:

Empty rack required for -RT parts

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Sterile
Color Natural
Model ART Non-filtered
Length (English) 3.5 in.
Length (Metric) 8.89 cm
Clean Claims DNase, RNase, DNA, ATP and Endotoxin Free
Format Rack
Tip Style Standard
Quantity 2800/Cs.
For Use With (Equipment) Universal Fit
Product Line ART
Fit for Purpose Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.