Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Artemether 98.0+%, TCI America™
SDP

Catalog No. A219025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A219025G 25 g
A21905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A219025G Supplier TCI America Supplier No. A219025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71963-77-4
Molecular Formula C16H26O5
Molecular Weight (g/mol) 298.379
MDL Number MFCD00866205
InChI Key SXYIRMFQILZOAM-ZSLLRAOYSA-N
PubChem CID 132451910
SMILES CC1CCC2C(C(OC3C24C1CCC(O3)(OO4)C)OC)C

Specifications

Melting Point 89°C
Color White
Quantity 25 g
Formula Weight 298.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Artemether
RTECSNumber KD4165000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.