Learn More
Selleck Chemical LLC Fulvestrant S1191-500mg
SDP
Supplier:  Selleck Chemical LLC S1191500MG
Encompass_Preferred
Fulvestrant is an estrogen receptor (ER) antagonist with IC50 of 0 94 nM in a cell-free assay Fulvestrant also induces autophagy and apoptosis and has antitumor activity
Catalog No. 50-310-8111
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.