Learn More
Selleck Chemical LLC Bumetanide S1287-50mg
SDP
Supplier:  Selleck Chemical LLC S128750MG
Encompass_Preferred
Bumetanide is a loop diuretic of the sulfamyl category to treat heart failure Bumetanide is a selective Na -K -Cl- cotransporter 1 (NKCC1) inhibitor weakly inhibits NKCC2 with IC50s of 0 68 and 4 0 M for hNKCC1A and hNKCC2A respectively
Catalog No. 50-311-1169
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.