Learn More
Medchemexpress LLC Artemisinin | 63968-64-9 | MFCD00081057 | 98.0% | 282.33 g/mol | C15H22O5 | 10 MG
Supplier: Medchemexpress LLC HYB0094R10MG
Artemisinin analytical standard supplied as a 10 mg reference material for research and analytical applications. This sesquiterpene lactone is used for assay calibration, method development, and analytical validation, and it has documented antimalarial activity and reported effects in cell-based studies. Provided with molecular formula C15H22O5 and molecular weight 282.33 g/mol.
- Intended for assay calibration and method development.
- Suitable for analytical validation and quality control workflows.
- Supplied in a 10 mg vial for convenient handling.
- Sesquiterpene lactone with molecular formula C15H22O5 and molecular weight 282.33 g/mol.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.