Learn More
ATCC Arthrobacter sp. ATCC 31651 Freeze-dried Culture
Supplier:  ATCC 31651
Encompass
Arthrobacter sp. (ATCC 31651) is a freeze-dried environmental bacterial strain supplied for laboratory research. The strain produces cholanic acid derivatives and is handled at biosafety level 1. It is intended for laboratory culture and study.
  • Freeze-dried format for stable, long-term storage.
  • Produces cholanic acid derivatives.
  • Biosafety level 1 organism suitable for standard laboratory handling.
  • Grows on peptone-sodium cholate medium at approximately 26 °C.
  • Intended for laboratory research use only.
Catalog No. NC3727260