Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Artist Brushes
SCI_ED

Catalog No. S30410
Change view
Click to view available options
Length (English):
7.75 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S30410 7.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30410 Supplier C&A Scientific Co Inc Supplier No. BR670/EMD

missing translation for 'validMainframeMsg'

Quality brushes with seamed metal ferrule and black polished handle.

  • Very useful for powders, reagents or Drosophila work
  • Overall length: 7.75 in. (19.7cm)

Specifications

Product Type Artist Brush
For Use With (Application) Very useful for powders, reagents or drosophila work
Includes Seamed metal ferrule and black polished handle
Length (English) 7.75 in.
Length (Metric) 19.7 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.