Learn More
Medchemexpress LLC HY-138642 25mg , ARV-471 CAS:2229711-68-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13864225MG
Encompass_Preferred
HY-138642 25mg ARV-471 CAS: 2229711-68-4 ARV-471 is an oral estrogen receptor PROTAC protein degrader for breast cancer. ARV-471 is a hetero-bifunctional molecule that facilitates the interactions between estrogen receptor alpha and an intracellular E3 ligase complex. ARV-471 leads to the ubiquitylation and subsequent degradation of estrogen receptors via the proteasome. ARV-471 robustly degrades ER in ER-positive breast cancer cell lines with a half-maximal degradation concentration (DC50) of ~2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0113
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.