Learn More
Selleck Chemical LLC ARV-771 1mg
SDP
Supplier:  Selleck Chemical LLC S85321MG
Encompass_Preferred
ARV-771 is a potentpan-(bromodomain and extra-terminal)BET degrader, a novel BET-PROTAC(proteolysis-targeting chimera) with Kd of 34 nM, 4.7 nM, 8.3 nM, 7.6 nM, 9.6 nM, and 7.6 nM for BRD2(1), BRD2(2), BRD3(1), BRD3(2), BRD4(1), and BRD4(2), respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3368
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.