Learn More
Enzo Life Sciences Arv-825 (25mg). CAS: 1818885-28-7
SDP
Supplier:  Enzo Life Sciences ENZCHM2580025
Encompass_Preferred
ARV-825 is a PROTAC targeting on BRD4, consisting of a BRD4-ligand JQ-1 linked to a cereblon E3 ligase ligand Pomalidomide, with Kds values of 90nM and 28nM for affinity to BD1 and BD2. Purity: ≥98%. Long Term Storage: -20°C.
Catalog No. 50-201-3715
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.