Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Arylsulfatase G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554729
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554729 Supplier Invitrogen™ Supplier No. PA554729
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YVTGIIGKWH LGHHGSYHPN FRGFDYYFGI PYSHDMGCTD TPGYNHPPCP ACPQGDGPSR NLQRDCYTDV ALPLYENLNI VEQPV Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.

Sulfatases, such as ARSG, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules. This gene, a novel arylsulfatase, has activity toward pseudosubstrates including p-nitrocatechol sulfate and 4-methylumbelliferyl sulfate. Activity is competitively inhibited by phosphate. The unprocessed protein is active as a 63-kDa monomer and demonstrates an acidic pH optimum as typically seen for lysosomal sulfatases. The protein accumulates within lysosomes and is also a glycoprotein that binds specifically to mannose 6-phosphate receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arylsulfatase G
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARSG
Gene Accession No. Q96EG1
Gene Alias 6330406P08Rik; AI846872; Arsg; Arylsulfatase G; ASG; KIAA1001; RGD1306571; UNQ839/PRO1777
Gene Symbols ARSG
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Arylsulfatase G. Recombinant protein control fragment (Product #RP-93287).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22901
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.