Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AS 1949490
GREENER_CHOICE

Catalog No. p-7116041
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-182-5 25 mg
37-185 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-182-5 Supplier Tocris Bioscience™ Supplier No. 3718/25
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

SH2 domain-containing inositol 5'-phosphatase 2 (SHIP2) inhibitor

AS 1949490 is a selective SHIP2 (SH2 domain-containing inositol 5'-phosphatase 2) inhibitor (IC50 values are 0.34 μM and 0.62 μM for mouse and human respectively); displays approximately 30-fold affinity for SHIP2 over SHIP1. Increases insulin-induced phosphorylation of Akt in L6 myotubules. Stimulates activation of glucose metabolism; regulates gluconeogenesis in vitro and in vivo and exhibits antidiabetic effects. Also promotes increased BDNF mRNA levels in cultured cortical neurons and has memory enhancing and antidepressant effects in vivo.

Specifications

Chemical Name or Material 3-[(4-Chlorophenyl)methoxy]-N-[(1S)-1-phenylethyl]thiophene-2-carboxamide
CAS 1203680-76-5
Assay Percent Range >99%
Quantity 25 mg
Molecular Formula C20H18ClNO2S
Formula Weight Observed MW: 371.88
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.