Learn More
Active Motif AS-8351
SDP
Supplier:  Active Motif 14112
Encompass_Preferred
Active Motif's AS-8351 has been used in a cocktail of nine small molecules to efficiently induce cardiac reprogramming of human fibroblasts. It is believed that AS-8351 exerts its effects by modulating the activity of the JmjC domain-containing histone demethylases (JmjC-KDM). Thus each of the 22 genes in the JmjC-KDM family was abrogated using small hairpin RNAs which showed that only knocking down KDM5B (or using another KDM5B inhibitor such as PBIT) could phenocopy AS-8351 in generating cardiomyocytes. This strongly suggests that KDM5B is the target of AS-8351.
Catalog No. NC1841016
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.