Learn More
Abcam AS601245 JNK Inhibitor, Selective, ATP Competitive, Solid, 5 mg
Description
- Potency: Potent, selective, ATP competitive JNK inhibitor (IC50 values are 150, 220 and 70 nM for hJNK1, 2 and 3 respectively)
- Physical Properties: MW 372.4 Da, Purity >95%, Solid form
- Biological Activity: Reduces cell adhesion and migration in cancer cells; shows neuroprotective and anti-ischemic effects in vivo
- Characteristics: Blood-brain barrier and cell-permeable
- Composition: 100% 2-(1,3-benzothiazol-2-yl)-2-[2-(2-pyridin-3-ylethylamino)pyrimidin-4-yl]acetonitrile
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | JNK (hJNK1, hJNK2, hJNK3) |
| Shipping Condition | Ambient |
| Buffer | 100% 2-(1,3-benzothiazol-2-yl)-2-[2-(2-pyridin-3-ylethylamino)pyrimidin-4-yl]acetonitrile |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Substrate Properties | Chemical Substrate |
| Formulation | 100% 2-(1,3-benzothiazol-2-yl)-2-[2-(2-pyridin-3-ylethylamino)pyrimidin-4-yl]acetonitrile |
| Quantity | 5 mg |
| For Use With (Application) | Cancer research, Neuroprotection, Anti-ischemic studies |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.