Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam AS601245, JNK Inhibitor, Selective, Solid, 1 mg

Catalog No. 502705411
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5411 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5411 Supplier Abcam Supplier No. AB1451941MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent, selective, and cell-permeable ATP-competitive JNK inhibitor.

  • Molecular weight: 372.4 Da
  • Purity: >95%
  • Form: Solid
  • Mechanism: Potent, selective, ATP competitive JNK inhibitor (IC50 values are 150, 220 and 70 nM for hJNK1, 2 and 3 respectively)
  • Key effects: Reduces cell adhesion and migration in cancer cells; shows neuroprotective and anti-ischemic effects in vivo
  • Properties: Blood-brain barrier and cell-permeable
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% 2-(1,3-benzothiazol-2-yl)-2-[2-(2-pyridin-3-ylethylamino)pyrimidin-4-yl]acetonitrile
Shipping Condition Ambient
Target Molecule JNK
Content And Storage Store at -20°C.
Form Solid
Formulation 100% 2-(1,3-benzothiazol-2-yl)-2-[2-(2-pyridin-3-ylethylamino)pyrimidin-4-yl]acetonitrile
Quantity 1 mg
For Use With (Application) Cancer research, Neuroscience research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.