Learn More
Invitrogen™ ASAH1 Polyclonal Antibody

Description
Immunogen sequence: ENSTSYEEAK NLLTKTKILA PAYFILGGNQ SGEGCVITRD RKESLDVYEL DAKQGRWYVV QTNYDRWKHP FFLDDRRTPA KMCLNRTSQE NISFETMYDV LSTKPVLNKL TVYTTLIDVT KGQF Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 86%.
Specifications
Specifications
| Antigen | ASAH1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ASAH1 |
| Gene Accession No. | Q13510 |
| Gene Alias | 2310081N20Rik; AC; ACDase; acid CDase; acid ceramidase; Acid ceramidase subunit alpha; Acid ceramidase subunit beta; Acylsphingosine deacylase; ASAH; Asah1; HSD33; HSD-33; N-acylethanolamine hydrolase ASAH1; N-acylsphingosine amidohydrolase; N-acylsphingosine amidohydrolase (acid ceramidase) 1; N-acylsphingosine amidohydrolase 1; PHP; PHP32; putative 32 kDa heart protein; SMAPME |
| Gene Symbols | ASAH1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.