Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASAP1 Polyclonal Antibody, Rockland™
SDP

Catalog No. p-7190979
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2129 100 μg
50-199-2130 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2129 Supplier Rockland Immunochemicals Supplier No. 600401GY1
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human ASAP1 protein. A BLAST analysis was used to suggest reactivity to human 100%, bovine 93%, and rat and mouse 87% homology for the immunogen sequence. Cross-reactivity from other sources has not been determined.

ASAP1, also known as Development and differentiation enhancing factor 1, is a protein with two ANK repeats, an Arf-GAP domain, a PH domain and a SH3 domain. ASAP1 is a homodimer known to interact with SRC and CRK and its activity is stimulated by phosphatidylinositol 4,5-biphosphate (PIP2). It possesses phosphatidylinositol 4,5-biphosphate-dependent GTPase-activating protein activity for ARF1 (ADP ribosylation factor 1) and ARF5 and a lesser activity towards ARF6. It may coordinate membrane trafficking with cell growth or actin cytoskeleton remodeling by binding to both SRC and PIP2 and may also function as a signal transduction protein involved in the differentiation of fibroblasts into adipocytes and possibly other cell types. It is widely expressed in most of the tissues. ASAP1 is a protooncogene involved in invasive breast cancer and in other types of invasive and malignant tumors, such as uveal melanomas.

Specifications

Antigen ASAP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene ASAP1
Gene Accession No. Q9ULH1
Gene Alias 130 kDa phosphatidylinositol 4,5-biphosphate-dependent ARF1 GTPase-activating protein; 130 kDa phosphatidylinositol 4,5-bisphosphate-dependent ARF1 GTPase-activating protein; ADP-ribosylation factor-directed GTPase-activating protein 1; AMAP1; ARF GTPase-activating protein 1; Arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 1; ArfGAP with SH3 domain, ankyrin repeat and PH domain 1; ArfGAP with SH3 domain, ankyrin repeat and PH domain1; ASAP1; AV239055; centaurin, beta 4; CENTB4; Ddef1; DEF-1; development and differentiation enhancing factor 1; Development and differentiation-enhancing factor 1; differentiation-enhancing factor 1; Kiaa1249; mKIAA1249; PAG2; PAP; PIP2-dependent ARF1 GAP; s19; Shag1; ZG14P
Gene Symbols ASAP1
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50807
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.