Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam ASB-14 Zwitterionic Detergent, Solid, 434.7 Da, 25 g

Catalog No. 502583328
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3328 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3328 Supplier Abcam Supplier No. AB14209525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zwitterionic detergent for solubilizing proteins for 2D analysis, including membrane proteins.

  • Molecular Weight: 434.7 Da
  • Purity: >97%
  • Form: Solid
  • Application: Solubilizes proteins for 2D analysis, including membrane proteins
  • Composition: 100% Propanaminium-1, N,N-Dimethyl-N-[3-[(1-oxotetradecyl)amino] propyl]-3-sulfo-, Inneres Salz

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Proteins
Shipping Condition Ambient
Buffer 100% Propanaminium-1, N,N-Dimethyl-N-[3-[(1-oxotetradecyl)amino] propyl]-3-sulfo-, inner salt
Content And Storage Store at 4°C.
Form Solid
Formulation 100% Propanaminium-1, N,N-Dimethyl-N-[3-[(1-oxotetradecyl)amino] propyl]-3-sulfo-, Inneres Salz
Quantity 25 g
For Use With (Application) Protein solubilization for 2D analysis, Membrane protein solubilization
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.