Learn More
Abcam ASB-14 Zwitterionic Detergent, Solid, 434.7 Da, 5 g
Description
- Molecular weight: 434.7 Da
- Purity: >97%
- Physical state: Solid
- Description: Zwitterionic detergent that solubilizes proteins, including membrane proteins, for 2D analysis
- Storage: Store at +4°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Proteins |
| Buffer | 100% Propanaminium-1, N,N-Dimethyl-N-[3-[(1-oxotetradecyl)amino] propyl]-3-sulfo-, inner salt |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Formulation | 100% Propanaminium-1, N,N-Dimethyl-N-[3-[(1-oxotetradecyl)amino] propyl]-3-sulfo-, Inneres Salz |
| Quantity | 5 g |
| For Use With (Application) | 2D analysis, protein solubilization, membrane protein analysis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.