Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Associated Bag Asbestos Hazard Bags
SDP

Catalog No. 19821940 Shop All Associated Bag Company Products
Change view
Click to view available options
Length (English):
24 in.
33 in.
38 in.
Height (English):
36 in.
40 in.
60 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Height (English)
19-821-940 38 in. 60 in.
19-821-941 33 in. 40 in.
19-821-942 24 in. 36 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-821-940 Supplier Associated Bag Supplier No. 658100C
Add to Cart
Add to Cart

Collect, transport, and dispose of asbestos-contaminated materials easily and safely

  • Printed with bright yellow warning message
  • Extra-thick, meets OSHA standard 1910.1001 for asbestos removal
  • Tamper-evident, self-locking nylon ties available

Specifications

Certifications/Compliance OSHA,1910.1001
Color Clear
For Use With (Application) Collect, transport and dispose of asbestos contaminated materials
Includes Biohazard label
Thickness (English) 6 mil
Length (English) 38 in.
Height (English) 60 in.
Product Type Asbestos Hazard Bag
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.