Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598420
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598420 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598420 Supplier Thermo Scientific Supplier No. PA598420
Only null left
Add to Cart
Add to Cart

ASC1 Polyclonal Antibody

ASC1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen ASC1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Trip4
Gene Accession No. Q15650
Gene Alias 4930558E03Rik; Activating signal cointegrator 1; Asc1; ASC-1; BB191711; HsT17391; SMABF1; thyroid hormone receptor interactor 4; thyroid receptor-interacting protein 4; transcription coactivator Asc1; TR-interacting protein 4; TRIP4; TRIP-4; ZC2HC5; zinc finger, C2HC5-type
Gene Symbols Trip4
Host Species Rabbit
Immunogen Recombinant Human Activating signal cointegrator 1 protein (360-535AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9325
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.