Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASC1 Recombinant Rabbit Monoclonal Antibody (PSH0-26)
GREENER_CHOICE

Catalog No. FEMA550797
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550797 Supplier Invitrogen™ Supplier No. MA550797
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: A549 cell lysate, Hela cell lysate, U87-MG cell lysate, 293T cell lysate, HL-60 cell lysate, HepG2 cell lysate, RH-35 cell lysate, rat small intestine tissue. Predicted band size: 66 kDa Subcellular Location: Nucleus, Cytoplasm, cytosol, Cytoplasm, cytoskeleton, microtubule organizing center, centrosome.

Transcription coactivator of nuclear receptors which functions in conjunction with CBP-p300 and SRC-1 and may play an important role in establishing distinct coactivator complexes under different cellular conditions. Plays a pivotal role in the transactivation of NF-kappa-B, SRF and AP1. Acts as a mediator of transrepression between nuclear receptor and either AP1 or NF-kappa-B. Plays a role in androgen receptor transactivation and in testicular function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone PSH0-26
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Trip4
Gene Accession No. Q15650
Gene Alias 4930558E03Rik; Activating signal cointegrator 1; Asc1; ASC-1; BB191711; HsT17391; SMABF1; thyroid hormone receptor interactor 4; thyroid receptor-interacting protein 4; transcription coactivator Asc1; TR-interacting protein 4; TRIP4; TRIP-4; ZC2HC5; zinc finger, C2HC5-type
Gene Symbols Trip4
Host Species Rabbit
Immunogen Recombinant protein within human ASC1 aa 1-581/581.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315769, 9325
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.