Learn More
Thomas Scientific ASCARITE II 8-20 MESH THO.500G UN1823 8 PG2
SDP
Supplier:  Thomas Scientific C049H40CS
Encompass_Preferred
Ascarite II CO2 Absorbent, 8-20 Mesh, 500 g, 12/CS.
Catalog No. NC9680658
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.