Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ascentis™ Express 160 Å Phenyl-Hexyl, 2.7 μm HPLC Column, Sold by MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Length (Metric):
3 cm
10 cm
15 cm
Description
- High-speed, high-performance liquid chromatography column based on a 160Å Fused-Core™ particle design
- The particle provides a thin porous shell of high-purity silica surrounding a solid silica core.
- This particle design exhibits very high column efficiency for high molecular weight solutes (up to 20 kDa) due to the shallow diffusion paths in the 0.5 μm thick porous shell and the small overall particle size of 2.7 μm.
- The endcapped, phenyl-hexyl bonded phase provides a stable, reversed phase packing with a pore structure and pore size that is optimized for reversed-phase HPLC separations of peptides and polypeptides, using typical acidic mobile phases favored for proteomic applications.

Specifications
Specifications
pH | 2 to 9 |
Particle Size | 2.7 μm |
Length (Metric) | 10 cm |
Pore Size | 160 Å pore size |
Surface Area | 90 m2/g |
Diameter (Metric) Inner | 2.1 mm |
For Use With (Equipment) | L11 Per USP |
Stationary Phase | Phenyl-Hexyl (dimethylphenyl-hexylsilane) bonding |
Endcapped | Endcapped |
Matrix | Fused-Core™ particle platform |
Ascentis is a registered trademark of Sigma-Aldrich Co. LLC, Fused-Core is a registered trademark of Advanced Materials Technology, Inc.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction