Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascomycin, 95%, Alfa Aesar™

Click to view available options
Quantity:
25mg
100mg
104987-12-4
C43H69NO12
792.02
MFCD06198665
ZDQSOHOQTUFQEM-NURRSENYSA-N
ascomycin,immunomycin,unii-auf4u5nsjk,ascomycin fk520,auf4u5nsjk,chembl8597,15,19-epoxy-3h-pyrido 2,1-c 1,4 oxaazacyclotricosine-1,7,20,21 4h,23h-tetrone, 8-ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-3-2-4-hydroxy-3-methoxycyclohexyl-1-methylethenyl-14,16-dimethoxy-4,10,12,18-tetram,ascomycin from streptomyces hygroscopicus var. ascomyceticus,15,19-epoxy-3h-pyrido 2,1-c 1,4 oxaazacyclotricosine-1,7,20,21 4h,23h-tetrone, 5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-8-ethyl-3-2-4-hydroxy-3-methoxycyclohexyl-1-methylethenyl-14,16-dimethoxy-4,10,12,18-tetramethyl-, 3s-3r* e 1s*,3s*,4s* ,4s*,19s*,26ar*,ascomycin fk 520, fr 900520, imm
5282071
CHEBI:29582
CCC1C=C(CC(CC(C2C(CC(C(O2)(C(=O)C(=O)N3CCCCC3C(=O)OC(C(C(CC1=O)O)C)C(=CC4CCC(C(C4)OC)O)C)O)C)OC)OC)C)C
104987-12-4
C43H69NO12
792.02
MFCD06198665
ZDQSOHOQTUFQEM-NURRSENYSA-N
ascomycin,immunomycin,unii-auf4u5nsjk,ascomycin fk520,auf4u5nsjk,chembl8597,15,19-epoxy-3h-pyrido 2,1-c 1,4 oxaazacyclotricosine-1,7,20,21 4h,23h-tetrone, 8-ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-3-2-4-hydroxy-3-methoxycyclohexyl-1-methylethenyl-14,16-dimethoxy-4,10,12,18-tetram,ascomycin from streptomyces hygroscopicus var. ascomyceticus,15,19-epoxy-3h-pyrido 2,1-c 1,4 oxaazacyclotricosine-1,7,20,21 4h,23h-tetrone, 5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-8-ethyl-3-2-4-hydroxy-3-methoxycyclohexyl-1-methylethenyl-14,16-dimethoxy-4,10,12,18-tetramethyl-, 3s-3r* e 1s*,3s*,4s* ,4s*,19s*,26ar*,ascomycin fk 520, fr 900520, imm
5282071
CHEBI:29582
CCC1C=C(CC(CC(C2C(CC(C(O2)(C(=O)C(=O)N3CCCCC3C(=O)OC(C(C(CC1=O)O)C)C(=CC4CCC(C(C4)OC)O)C)O)C)OC)OC)C)C

Internal standard for LC-MS/MS testing of the immunosuppressants tacrolimus, sirolimus, and everolimus.

Applications
Ascomycin is a potent immunosuppressive agent that could be used as a potential therapeutic agent for autoimmune diseases. Ascomycin is an inhibitor of PP2. It is an internal standard for LC-MS/MS testing of the immunosuppressants tacrolimus, sirolimus, and everolimus. This Certified Spiking Solution™ is suitable for critical quantitative applications in clinical and diagnostic testing such as therapeutic drug monitoring assays to ensure patients remain within the narrow therapeutic range of these immunosuppressants.

Solubility
Soluble in DMSO at 65mg/ml; soluble in ethanol at 50mg/m; with warming. Very poorly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Quantity 25mg
Flash Point >110°C(230°F)
Color White
Boiling Point 868°C
Melting Point 156°C to 160°C
Solubility Information Soluble in DMSO at 65mg/mL; soluble in ethanol at 50mg/mL; with warming. Very poorly soluble in water
Formula Weight 790
Physical Form Powder
Percent Purity 95%
Chemical Name or Material Ascomycin

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : KD4185000

storageNote1 : −30° to −10°C