Learn More
Apexbio Technology LLC Ascomycin(FK 520) 104987-12-4 50mg
SDP
Supplier:  Apexbio Technology LLC A319150
Encompass_Preferred
Ascomycin FK 520 (CAS 104987-12-4) is a small-molecule immunosuppressant targeting calcineurin phosphatase via binding to immunophilin FKBP12 It is designed to inhibit calcineurin activity thereby suppressing NFAT-mediated T-cell activation and proliferation Ascomycin FK 520 exerts its biological activity primarily by forming a complex with FKBP12 that inhibits calcineurin phosphatase In in vitro assays Ascomycin FK 520 demonstrates dose-dependent inhibition of lymphocyte responsiveness with an IC50 in the submicromolar range and no cytotoxicity observed below 3 2 M In animal models administration at doses 3 2 mg/kg prolongs skin transplant survival in rats Based on these pharmacological properties Ascomycin FK 520 holds research potential in studies of calcineurin-mediated immune responses and the development of T-cell modulation strategies
Catalog No. 50-000-95198
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.